Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC46A1 Full-length ORF (ADZ16056.1) Recombinant Protein without tag

Catalog No. 89962361 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-361 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-361 Supplier Abnova™ Supplier No. H00113235G01
Add to Cart
Add to Cart

Used for AP, Func, Screening

This gene encodes a transmembrane proton-coupled folate transporter protein that facilitates the movement of folate and antifolate substrates across cell membranes optimally in acidic pH environments. This protein is also expressed in the brain and choroid plexus where it transports folates into the central nervous system. This protein further functions as a transmembrane heme transporter in duodenal enterocytes and, potentially, in other tissues like liver and kidney. Its localization to the apical membrane or cytoplasm of intestinal cells is modulated by dietary iron levels. Mutations in this gene cause the autosomal recessive hereditary folate malabsorption (HFM) disease. HFM is characterized by folate deficiency due to reduced intestinal folate absorption and subsequent anemia, hypoimmunoglobulinemia, and recurrent infections. [provided by RefSeq]

Sequence: MEGSASPPEKPRARPAAAVLCRGPVEPLVFLANFALVLQGPLTTQYLWHRFSADLGYNGTRQRGGCSNRSADPTMQEVETLTSHWTLYMNVGGFLVGLFSSTLLGAWSDSVGRRPLLVLASLGLLLQALVSVFVVQLQLHVGYFVLGRILCALLGDFGGLLAASFASVADVSSSRSRTFRMALLEASIGVAGMLASLLGGHWLRAQGYANPFWLALALLIAMTLYAAFCFGETLKEPKSTRLFTFRHHRSIVQLYVAPAPEKSRKHLALYSLAIFVVITVHFGAQDILTLYELSTPLCWDSKLIGYGSAAQHLPYLTSLLALKLLQYCLADAWVAEIGLAFNILGMVVFAFATITPLMFTGALFSAVACVNSLAMLTASGIFNSLYPATLNFMKGFPFLLGAGLLLIPAVLIGMLEKADPHLEFQQFPQSP

Specifications

Accession Number ADZ16056.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 113235
Molecular Weight (g/mol) 47.5kDa
Name SLC46A1 (Human) Recombinant Protein
Quantity 10 μg
Immunogen MEGSASPPEKPRARPAAAVLCRGPVEPLVFLANFALVLQGPLTTQYLWHRFSADLGYNGTRQRGGCSNRSADPTMQEVETLTSHWTLYMNVGGFLVGLFSSTLLGAWSDSVGRRPLLVLASLGLLLQALVSVFVVQLQLHVGYFVLGRILCALLGDFGGLLAASFASVADVSSSRSRTFRMALLEASIGVAGMLASLLGGHWLRAQGYANPFWLALALLIAMTLYAAFCFGETLKEPKSTRLFTFRHHRSIVQLYVAPAPEKSRKHLALYSLAIFVVITVHFGAQDILTLYELSTPLCWDSKLIGYGSAAQHLPYLTSLLALKLLQYCLADAWVAEIGLAFNILGMVVFAFATITPLMFTGALFSAVACVNSLAMLTASGIFNSLYPATLNFMKGFPFLLGAGLLLIPAVLIGMLEKADPHLEFQQFPQSP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HCP1/MGC9564/PCFT
Common Name SLC46A1
Gene Symbol SLC46A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.