Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC7A10 Full-length ORF (ADZ15434.1) Recombinant Protein without tag

Catalog No. 89962412 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-412 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-412 Supplier Abnova™ Supplier No. H00056301G01
Add to Cart
Add to Cart

Used for AP, Func, Screening

SLC7A10, in association with 4F2HC (SLC3A2; MIM 158070), mediates high-affinity transport of D-serine and several other neutral amino acids (Nakauchi et al., 2000 [PubMed 10863037]).[supplied by OMIM]

Sequence: MAGHTQQPSGRGNPRPAPSPSPVPGTVPGASERVALKKEIGLLSACTIIIGNIIGSGIFISPKGVLEHSGSVGLALFVWVLGGGVTALGSLCYAELGVAIPKSGGDYAYVTEIFGGLAGFLLLWSAVLIMYPTSLAVISMTFSNYVLQPVFPNCIPPTTASRVLSMACLMLLTWVNSSSVRWATRIQDMFTGGKLLALSLIIGVGLLQIFQGHFEELRPSNAFAFWMTPSVGHLALAFLQGSFAFSGWNFLNYVTEEMVDARKNLPRAIFISIPLVTFVYTFTNIAYFTAMSPQELLSSNAVAVTFGEKLLGYFSWVMPVSVALSTFGGINGYLFTYSRLCFSGAREGHLPSLLAMIHVRHCTPIPALLVCCGATAVIMLVGDTYTLINYVSFINYLCYGVTILGLLLLRWRRPALHRPIKVNLLIPVAYLVFWAFLLVFSFISEPMVCGVGVIIILTGVPIFFLGVFWRSKPKCVHRLTESMTHWGQELCFVVYPQDAPEEEENGPCPPSLLPATDKPSKPQ

Specifications

Accession Number ADZ15434.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 56301
Molecular Weight (g/mol) 57.6kDa
Name SLC7A10 (Human) Recombinant Protein
Quantity 10 μg
Immunogen MAGHTQQPSGRGNPRPAPSPSPVPGTVPGASERVALKKEIGLLSACTIIIGNIIGSGIFISPKGVLEHSGSVGLALFVWVLGGGVTALGSLCYAELGVAIPKSGGDYAYVTEIFGGLAGFLLLWSAVLIMYPTSLAVISMTFSNYVLQPVFPNCIPPTTASRVLSMACLMLLTWVNSSSVRWATRIQDMFTGGKLLALSLIIGVGLLQIFQGHFEELRPSNAFAFWMTPSVGHLALAFLQGSFAFSGWNFLNYVTEEMVDARKNLPRAIFISIPLVTFVYTFTNIAYFTAMSPQELLSSNAVAVTFGEKLLGYFSWVMPVSVALSTFGGINGYLFTYSRLCFSGAREGHLPSLLAMIHVRHCTPIPALLVCCGATAVIMLVGDTYTLINYVSFINYLCYGVTILGLLLLRWRRPALHRPIKVNLLIPVAYLVFWAFLLVFSFISEPMVCGVGVIIILTGVPIFFLGVFWRSKPKCVHRLTESMTHWGQELCFVVYPQDAPEEEENGPCPPSLLPATDKPSKPQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ20839/HASC-1/asc-1
Common Name SLC7A10
Gene Symbol SLC7A10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.