Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC7A7 Partial ORF (NP_003973.2, 462 a.a. - 511 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962422 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-422 25 μg
89-962-421 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-422 Supplier Abnova™ Supplier No. H00009056Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is the light subunit of a cationic amino acid transporter. This sodium-independent transporter is formed when the light subunit encoded by this gene dimerizes with the heavy subunit transporter protein SLC3A2. This transporter is found in epithelial cell membranes where it transfers cationic and large neutral amino acids from the cell to the extracellular space. Defects in this gene are a cause of lysinuric protein intolerance (LPI). Several transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: RVPEHKRPLYLRRIVGSATRYLQVLCMSVAAEMDLEDGGEMPKQRDPKSN

Specifications

Accession Number NP_003973.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9056
Molecular Weight (g/mol) 31.24kDa
Name SLC7A7 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen RVPEHKRPLYLRRIVGSATRYLQVLCMSVAAEMDLEDGGEMPKQRDPKSN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LAT3/LPI/MOP-2/Y+LAT1/y+LAT-1
Common Name SLC7A7
Gene Symbol SLC7A7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.