Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC8A3 Full-length ORF (NP_891981.1, 1 a.a. - 284 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-430 10 ug
89-962-431 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-430 Supplier Abnova™ Supplier No. H00006547P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the sodium/calcium exchanger integral membrane protein family. Three mammalian isoforms in family 8 have been identified. Na+/Ca2+ exchange proteins are involved in maintaining Ca2+ homeostasis in a wide variety of cell types. The protein is regulated by intracellular calcium ions and is found in both the plasma membrane and intracellular organellar membranes, where exchange of Na+ for Ca2+ occurs in an electrogenic manner. Alternative splicing has been observed for this gene and multiple variants have been described. [provided by RefSeq]

Sequence: MEEEEAKRIAEMGKPVLGEHPKLEVIIEESYEFKTTVDKLIKKTNLALVVGTHSWRDQFMEAITVSAAGDEDEDESGEERLPSCFDYVMHFLTVFWKVLFACVPPTEYCHGWACFAVSILIIGMLTAIIGDLASHFGCTIGLKDSVTAVVFVAFGTSVPDTFASKAAALQDVYADASIGNVTGSNAVNVFLGIGLAWSVAAIYWALQGQEFHVSAGTLAFSVTLFTIFAFVCISVLLYRRRPHLGGELGGPRGCKLATTWLFVSLWLLYILFATLEAYCYIKGF

Specifications

Accession Number NP_891981.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6547
Molecular Weight (g/mol) 57.5kDa
Name SLC8A3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MEEEEAKRIAEMGKPVLGEHPKLEVIIEESYEFKTTVDKLIKKTNLALVVGTHSWRDQFMEAITVSAAGDEDEDESGEERLPSCFDYVMHFLTVFWKVLFACVPPTEYCHGWACFAVSILIIGMLTAIIGDLASHFGCTIGLKDSVTAVVFVAFGTSVPDTFASKAAALQDVYADASIGNVTGSNAVNVFLGIGLAWSVAAIYWALQGQEFHVSAGTLAFSVTLFTIFAFVCISVLLYRRRPHLGGELGGPRGCKLATTWLFVSLWLLYILFATLEAYCYIKGF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias NCX3
Common Name SLC8A3
Gene Symbol SLC8A3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.