Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC9A1 Full-length ORF (ADZ15465.1) Recombinant Protein without tag

Catalog No. 89962434 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-434 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-434 Supplier Abnova™ Supplier No. H00006548G01
Add to Cart
Add to Cart

Used for AP, Func, Screening

The Na+/H+ antiporter (SLC9A1) is a ubiquitous membrane-bound enzyme involved in pH regulation of vertebrate cells. It is specifically inhibited by the diuretic drug amiloride and activated by a variety of signals including growth factors, mitogens, neurotransmitters, tumor promoters, and others (Mattei et al., 1988 [PubMed 2846238]).[supplied by OMIM]

Sequence: MVLRSGICGLSPHRIFPSLLVVVALVGLLPVLRSHGLQLSPTASTIRSSEPPRERSIGDVTTAPPEVTPESRPVNHSVTDHGMKPRKAFPVLGIDYTHVRTPFEISLWILLACLMKIGFHVIPTISSIVPESCLLIVVGLLVGGLIKGVGETPPFLQSDVFFLFLLPPIILDAGYFLPLRQFTENLGTILIFAVVGTLWNAFFLGGLMYAVCLVGGEQINNIGLLDNLLFGSIISAVDPVAVLAVFEEIHINELLHILVFGESLLNDAVTVVLYHLFEEFANYEHVGIVDIFLGFLSFFVVALGGVLVGVVYGVIAAFTSRFTSHIRVIEPLFVFLYSYMAYLSAELFHLSGIMALIASGVVMRPYVEANISHKSHTTIKYFLKMWSSVSETLIFIFLGVSTVAGSHHWNWTFVISTLLFCLIARVLGVLGLTWFINKFRIVKLTPKDQFIIAYGGLRGAIAFSLGYLLDKKHFPMCDLFLTAIITVIFFTVFVQVLGQGRAGPCLGDPHRLFPWKERKACDLKCDSSPSSTTNLLCDLGRATPPFWASVSSIVK

Specifications

Accession Number ADZ15465.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 6548
Molecular Weight (g/mol) 61.1kDa
Name SLC9A1 (Human) Recombinant Protein
Quantity 10 μg
Immunogen MVLRSGICGLSPHRIFPSLLVVVALVGLLPVLRSHGLQLSPTASTIRSSEPPRERSIGDVTTAPPEVTPESRPVNHSVTDHGMKPRKAFPVLGIDYTHVRTPFEISLWILLACLMKIGFHVIPTISSIVPESCLLIVVGLLVGGLIKGVGETPPFLQSDVFFLFLLPPIILDAGYFLPLRQFTENLGTILIFAVVGTLWNAFFLGGLMYAVCLVGGEQINNIGLLDNLLFGSIISAVDPVAVLAVFEEIHINELLHILVFGESLLNDAVTVVLYHLFEEFANYEHVGIVDIFLGFLSFFVVALGGVLVGVVYGVIAAFTSRFTSHIRVIEPLFVFLYSYMAYLSAELFHLSGIMALIASGVVMRPYVEANISHKSHTTIKYFLKMWSSVSETLIFIFLGVSTVAGSHHWNWTFVISTLLFCLIARVLGVLGLTWFINKFRIVKLTPKDQFIIAYGGLRGAIAFSLGYLLDKKHFPMCDLFLTAIITVIFFTVFVQVLGQGRAGPCLGDPHRLFPWKERKACDLKCDSSPSSTTNLLCDLGRATPPFWASVSSIVK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias APNH/FLJ42224/NHE1
Common Name SLC9A1
Gene Symbol SLC9A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.