Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC9A3R1 Partial ORF (NP_004243.1, 252 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962438 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-438 25 μg
89-962-437 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-438 Supplier Abnova™ Supplier No. H00009368Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a sodium/hydrogen exchanger regulatory cofactor. The protein interacts with and regulates various proteins including the cystic fibrosis transmembrane conductance regulator and G-protein coupled receptors such as the beta2-adrenergic receptor and the parathyroid hormone 1 receptor. The protein also interacts with proteins that function as linkers between integral membrane and cytoskeletal proteins. The protein localizes to actin-rich structures including membrane ruffles, microvilli, and filopodia. Mutations in this gene result in hypophosphatemic nephrolithiasis/osteoporosis type 2, and loss of heterozygosity of this gene is implicated in breast cancer

Sequence: NGPLPVPFTNGEIQKENSREALAEAALESPRPALVRSASSDTSEELNSQDSPPKQDSTAPSSTSSSDPILDFNISLAMAKERAHQKRSSKRAPQMDWSKKNELFSNL

Specifications

Accession Number NP_004243.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9368
Molecular Weight (g/mol) 37.51kDa
Name SLC9A3R1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen NGPLPVPFTNGEIQKENSREALAEAALESPRPALVRSASSDTSEELNSQDSPPKQDSTAPSSTSSSDPILDFNISLAMAKERAHQKRSSKRAPQMDWSKKNELFSNL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EBP50/NHERF/NHERF1/NPHLOP2
Common Name SLC9A3R1
Gene Symbol SLC9A3R1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.