Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC9A6 Full-length ORF (NP_001036002.1, 1 a.a. - 701 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962444 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-444 10 μg
89-962-445 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-444 Supplier Abnova™ Supplier No. H00010479P01S

Used for AP, Array, ELISA, WB-Re

The SLC9A6 gene encodes a monovalent sodium-selective sodium/hydrogen exchanger (NHE) that is found in the membranes of intracellular organelles such as mitochondria and endosomes. NHEs participate in a wide array of essential cellular processes, including control of intracellular pH, maintenance of cellular volume, and reabsorption of sodium across renal, intestinal, and other epithelia.[supplied by OMIM]

Sequence: MARRGWRRAPLRRGVGSSPRARRLMRPLWLLLAVGVFDWAGASDGGGGEARAMDEEIVSEKQAEESHRQDSANLLIFILLLTLTILTIWLFKHRRARFLHETGLAMIYGLLVGLVLRYGIHVPSDVNNVTLSCEVQSSPTTLLVNVSGKFYEYMLKGEISSHELNNVQDNEMLRKVTFDPEVFFNILLPPIIFYAGYSLKRRHFFRNLGSILAYAFLGTAISCFVIGSIMYGCVTLMKVTGQLAGDFYFTDCLLFGAIVSATDPVTVLAIFHELQVDVELYALLFGESVLNDAVAIVLSSSIVAYQPAGDNSHTFDVTAMFKSIGIFLGIFSGSFAMGAATGVVTALVTKFTKLREFQLLETGLFFLMSWSTFLLAEAWGFTGVVAVLFCGITQAHYTYNNLSTESQHRTKQLFELLNFLAENFIFSYMGLTLFTFQNHVFNPTFVVGAFVAIFLGRAANIYPLSLLLNLGRRSKIGSNFQHMMMFAGLRGAMAFALAIRDTATYARQMMFSTTLLIVFFTVWVFGGGTTAMLSCLHIRVGVDSDQEHLGVPENERRTTKAESAWLFRMWYNFDHNYLKPLLTHSGPPLTTTLPACCGPIARCLTSPQAYENQEQLKDDDSDLILNDGDISLTYGDSTVNTEPATSSAPRRFMGNSSEDALDRELAFGDHELVIRGTRLVLPMDDSEPPLNLLDNTRHGPA

Specifications

Accession Number NP_001036002.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10479
Molecular Weight (g/mol) 104.3kDa
Name SLC9A6 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MARRGWRRAPLRRGVGSSPRARRLMRPLWLLLAVGVFDWAGASDGGGGEARAMDEEIVSEKQAEESHRQDSANLLIFILLLTLTILTIWLFKHRRARFLHETGLAMIYGLLVGLVLRYGIHVPSDVNNVTLSCEVQSSPTTLLVNVSGKFYEYMLKGEISSHELNNVQDNEMLRKVTFDPEVFFNILLPPIIFYAGYSLKRRHFFRNLGSILAYAFLGTAISCFVIGSIMYGCVTLMKVTGQLAGDFYFTDCLLFGAIVSATDPVTVLAIFHELQVDVELYALLFGESVLNDAVAIVLSSSIVAYQPAGDNSHTFDVTAMFKSIGIFLGIFSGSFAMGAATGVVTALVTKFTKLREFQLLETGLFFLMSWSTFLLAEAWGFTGVVAVLFCGITQAHYTYNNLSTESQHRTKQLFELLNFLAENFIFSYMGLTLFTFQNHVFNPTFVVGAFVAIFLGRAANIYPLSLLLNLGRRSKIGSNFQHMMMFAGLRGAMAFALAIRDTATYARQMMFSTTLLIVFFTVWVFGGGTTAMLSCLHIRVGVDSDQEHLGVPENERRTTKAESAWLFRMWYNFDHNYLKPLLTHSGPPLTTTLPACCGPIARCLTSPQAYENQEQLKDDDSDLILNDGDISLTYGDSTVNTEPATSSAPRRFMGNSSEDALDRELAFGDHELVIRGTRLVLPMDDSEPPLNLLDNTRHGPA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0267/MRSA/NHE6
Common Name SLC9A6
Gene Symbol SLC9A6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.