Learn More
Abcam Human Smad2 Phospho S465 and S467 Peptide, 1 mg
Description
- Target Protein: Smad2
- Protein Type: Synthetic peptide
- Purity: >90%
- Species Reactivity: Human
- Form: Lyophilized
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Smad2 (Phospho S465 + S467) |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Label or Dye | Unconjugated |
| Shipping Condition | Blue Ice |
| Cross Reactivity | Human |
| Quantity | 1 mg |
| For Use With (Application) | Blocking, Control |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.