Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SMARCD3 Full-length ORF (NP_001003802.1, 1 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962552 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-552 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-552 Supplier Abnova™ Supplier No. H00006604P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined. [provided by RefSeq]

Sequence: MTPGLQHPPTVVQRPGMPSGARMPHQGAPMGPPGSPYMGSPAVRPGLAPAGMEPARKRAAPPPGQSQAQSQGQPVPTAPARSRSAKRRKMADKILPQRIRELVPESQAYMDLLAFERKLDQTIMRKRVDIQEALKRPMKQKRKLRLYISNTFNPAKPDAEDSDGSIASWELRVEGKLLDDPSKQKRKFSSFFKSLVIELDKDLYGPDNHLVEWHRTPTTQETDGFQVKRPGDLSVRCTLLLMLDYQPPQFKLDPRLARLLGLHTQSRSAIVQALWQYVKTNRLQDSHDKEYINGDKYFQQIFDCPRLKFSEIPQRLTALLLPPDPIVINHVISVDPSDQKKTACYDIDVEVEEPLKGQMSSFLLSTANQQEISALDSKIHETIESINQLKIQRDFMLSFSRDPKGYVQDLLRSQSRDLKVMTDVAGNPEEERRAEFYHQPWSQEAVSRYFYCKIQQRRQELEQSLVVRNT

Specifications

Accession Number NP_001003802.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6604
Molecular Weight (g/mol) 80.1kDa
Name SMARCD3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MTPGLQHPPTVVQRPGMPSGARMPHQGAPMGPPGSPYMGSPAVRPGLAPAGMEPARKRAAPPPGQSQAQSQGQPVPTAPARSRSAKRRKMADKILPQRIRELVPESQAYMDLLAFERKLDQTIMRKRVDIQEALKRPMKQKRKLRLYISNTFNPAKPDAEDSDGSIASWELRVEGKLLDDPSKQKRKFSSFFKSLVIELDKDLYGPDNHLVEWHRTPTTQETDGFQVKRPGDLSVRCTLLLMLDYQPPQFKLDPRLARLLGLHTQSRSAIVQALWQYVKTNRLQDSHDKEYINGDKYFQQIFDCPRLKFSEIPQRLTALLLPPDPIVINHVISVDPSDQKKTACYDIDVEVEEPLKGQMSSFLLSTANQQEISALDSKIHETIESINQLKIQRDFMLSFSRDPKGYVQDLLRSQSRDLKVMTDVAGNPEEERRAEFYHQPWSQEAVSRYFYCKIQQRRQELEQSLVVRNT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BAF60C/CRACD3/MGC111010/Rsc6p
Common Name SMARCD3
Gene Symbol SMARCD3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.