Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SMARCE1 Full-length ORF (AAH07082.1, 1 a.a. - 411 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962555 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-555 10 ug
89-962-556 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-555 Supplier Abnova™ Supplier No. H00006605P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is part of the large ATP-dependent chromatin remodeling complex SWI/SNF, which is required for transcriptional activation of genes normally repressed by chromatin. The encoded protein, either alone or when in the SWI/SNF complex, can bind to 4-way junction DNA, which is thought to mimic the topology of DNA as it enters or exits the nucleosome. The protein contains a DNA-binding HMG domain, but disruption of this domain does not abolish the DNA-binding or nucleosome-displacement activities of the SWI/SNF complex. Unlike most of the SWI/SNF complex proteins, this protein has no yeast counterpart. [provided by RefSeq]

Sequence: MSKRPSYAPPPTPAPATQMPSTPGFVGYNPYSHLAYNNYRLGGNPSTNSRVTASSGITIPKPPKPPDKPLMPYMRYSRKVWDQVKASNPDLKLWEIGKIIGGMWRDLTDEEKQEYLNEYEAEKIEYNESMKAYHNSPAYLAYINAKSRAEAALEEESRQRQSRMEKGEPYMSIQPAEDPDDYDDGFSMKHTATARFQRNHRLISEILSESVVPDVRSVVTTARMQVLKRQVQSLMVHQRKLEAELLQIEERHQEKKRKFLESTDSFNNELKRLCGLKVEVDMEKIAAEIAQAEEQARKRQEEREKEAAEQAERSQSSIVPEEEQAANKGEEKKDDENIPMETEETHLEETTESQQNGEEGTSTPEDKESGQEGVDSMAEEGTSDSNTGSESNSATVEEPPTDPIPEDEKKE

Specifications

Accession Number AAH07082.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6605
Molecular Weight (g/mol) 73.1kDa
Name SMARCE1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MSKRPSYAPPPTPAPATQMPSTPGFVGYNPYSHLAYNNYRLGGNPSTNSRVTASSGITIPKPPKPPDKPLMPYMRYSRKVWDQVKASNPDLKLWEIGKIIGGMWRDLTDEEKQEYLNEYEAEKIEYNESMKAYHNSPAYLAYINAKSRAEAALEEESRQRQSRMEKGEPYMSIQPAEDPDDYDDGFSMKHTATARFQRNHRLISEILSESVVPDVRSVVTTARMQVLKRQVQSLMVHQRKLEAELLQIEERHQEKKRKFLESTDSFNNELKRLCGLKVEVDMEKIAAEIAQAEEQARKRQEEREKEAAEQAERSQSSIVPEEEQAANKGEEKKDDENIPMETEETHLEETTESQQNGEEGTSTPEDKESGQEGVDSMAEEGTSDSNTGSESNSATVEEPPTDPIPEDEKKE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BAF57
Common Name SMARCE1
Gene Symbol SMARCE1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.