Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SMPD1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7066414
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5348SP 25 μg
MAB5348 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5348SP Supplier R&D Systems Supplier No. MAB5348SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SMPD1 Monoclonal specifically detects SMPD1 in Human samples. It is validated for Immunoprecipitation, Neutralization.

Specifications

Antigen SMPD1
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 563418
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 25 ug/mL, Neutralization 0.384 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_000534
Gene Alias Acid sphingomyelinase, ASMASE, ASMsphingomyelin phosphodiesterase, EC 3.1.4.12, NPD, sphingomyelin phosphodiesterase 1, acid lysosomal
Gene Symbols SMPD1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SMPD1 isoform 1 His62-Pro628 Accession # NP_000534
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6609
Test Specificity Detects human SMPD1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human SMPD3 and recombinant mouse SMPD1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.