Learn More
Description
- Also known as: ASM3A; ASML3a; yR36GH4.1
- SMPDL3A belongs to the acid sphingomyelinase family. It is detected in blood serum and macrophages.
- SMPDL3A is a glycosylated protein that is upregulated in macrophages in response to cholesterol accumulation.
- It has phosphodiesterase activity with nucleoside triphosphates as preferred substrates.
- The SMPDL3A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hSMPDL3A |
| Accession Number | NP_006705 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1375 bp |
| Gene ID (Entrez) | 10924 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
