Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SNCAIP Full-length ORF (AAH94759.1, 1 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962684 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-684 10 μg
89-962-685 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-684 Supplier Abnova™ Supplier No. H00009627P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein containing several protein-protein interaction domains, including ankyrin-like repeats, a coiled-coil domain, and an ATP/GTP-binding motif. The encoded protein interacts with alpha-synuclein in neuronal tissue and may play a role in the formation of cytoplasmic inclusions and neurodegeneration. A mutation in this gene has been associated with Parkinson's disease. Alternatively spliced transcript variants encoding different isoforms of this gene have been described, but their full-length nature has yet to be determined. [provided by RefSeq]

Sequence: MTYLIQSHHSRRSQNCAEDVIRKTKTDQNGQLECVRWMVSETEAIAELSCSKDFPSLIHYAGCYGQEKILLWLLQFMQEQGISLDEVDQDGNSAVHVASQHGYLGCIQTLVEYGANVTMQNHAGEKPSQSAERQGHTLCSRYLVVVETCMSLASQVVKLTKQLKEQTVERVTLQNQLQQFLEAQKSEGKSLPSSPSSPSSPASRKSQWKSPDADDDSVAKSKPGVQEGIQVLGSLSASSRARPKAKDEDSDKILRQLLGKEISENVCTQEKLSLEFQDAQASSRNSKKIPLEKRELKLARLRQLMQRSLSESDTDSNNSEDPKTTPVRKADRPRPQPIVESVESMGSAESLHLMIKKHTLASGGRRFPFSIKASKSLDGHSPSPTSESSEPDLESQYPGSGSIPPNQPSGDPQQPSPDSTAAQKVATSPKSALKSPSSKRRTSQNLKLRVTFEEPVVQMEQPSLELNGEKDKDKGRTLQRTSTSNESGDQLKRPFGAFRSIMETLSGNQNNNNNYQAANQLKTSTLPLTSLGRKTDAKGNPASSASKGKNKAEMYSSCINLSSNMLIEEHLCNDTRHNDINRKMKKSYSIKHIAEPESKELFL

Specifications

Accession Number AAH94759.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9627
Molecular Weight (g/mol) 93kDa
Name SNCAIP (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MTYLIQSHHSRRSQNCAEDVIRKTKTDQNGQLECVRWMVSETEAIAELSCSKDFPSLIHYAGCYGQEKILLWLLQFMQEQGISLDEVDQDGNSAVHVASQHGYLGCIQTLVEYGANVTMQNHAGEKPSQSAERQGHTLCSRYLVVVETCMSLASQVVKLTKQLKEQTVERVTLQNQLQQFLEAQKSEGKSLPSSPSSPSSPASRKSQWKSPDADDDSVAKSKPGVQEGIQVLGSLSASSRARPKAKDEDSDKILRQLLGKEISENVCTQEKLSLEFQDAQASSRNSKKIPLEKRELKLARLRQLMQRSLSESDTDSNNSEDPKTTPVRKADRPRPQPIVESVESMGSAESLHLMIKKHTLASGGRRFPFSIKASKSLDGHSPSPTSESSEPDLESQYPGSGSIPPNQPSGDPQQPSPDSTAAQKVATSPKSALKSPSSKRRTSQNLKLRVTFEEPVVQMEQPSLELNGEKDKDKGRTLQRTSTSNESGDQLKRPFGAFRSIMETLSGNQNNNNNYQAANQLKTSTLPLTSLGRKTDAKGNPASSASKGKNKAEMYSSCINLSSNMLIEEHLCNDTRHNDINRKMKKSYSIKHIAEPESKELFL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC39814/SYPH1
Common Name SNCAIP
Gene Symbol SNCAIP
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.