Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SNRNP35 Full-length ORF (NP_851030.1, 1 a.a. - 251 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966307 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-307 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-307 Supplier Abnova™ Supplier No. H00011066P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a homolog of U1-snRNP binding protein. The N-terminal half contains a RNA recognition motif and the C-terminal is rich in Arg/Asp and Arg/Glu dipeptides; a characteristic of a variety of splicing factors. This protein is a component of the U11/U12 small nuclear ribonucleoproteins (snRNP) that form part of the U12-type spliceosome. This gene is differentially expressed in a variety of human tissues. Alternative splicing results in multiple transcript variants encoding distinct proteins

Sequence: MKPANMNDWMPIAKEYDPLKAGSIDGTDEDPHDRAVWRAMLARYVPNKGVIGDPLLTLFVARLNLQTKEDKLKEVFSRYGDIRRLRLVRDLVTGFSKGYAFIEYKEERAVIKAYRDADGLVIDQHEIFVDYELERTLKGWIPRRLGGGLGGKKESGQLRFGGRDRPFRKPINLPVVKNDLYREGKRERRERSRSRERHWDSRTRDRDHDRGREKRWQEREPTRVWPDNDWERERDFRDDRIKGREKKERGK

Specifications

Accession Number NP_851030.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11066
Molecular Weight (g/mol) 56.4kDa
Name SNRNP35 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MKPANMNDWMPIAKEYDPLKAGSIDGTDEDPHDRAVWRAMLARYVPNKGVIGDPLLTLFVARLNLQTKEDKLKEVFSRYGDIRRLRLVRDLVTGFSKGYAFIEYKEERAVIKAYRDADGLVIDQHEIFVDYELERTLKGWIPRRLGGGLGGKKESGQLRFGGRDRPFRKPINLPVVKNDLYREGKRERRERSRSRERHWDSRTRDRDHDRGREKRWQEREPTRVWPDNDWERERDFRDDRIKGREKKERGK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HM-1/MGC138160/U1SNRNPBP
Common Name SNRNP35
Gene Symbol SNRNP35
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.