Learn More
Invitrogen™ Human SNX1 (aa 139-155) Synthetic Peptide
Description
Human SNX1 (residues 139-155) Synthetic Peptide, PEP-286, from Invitrogen.
Specifications
Specifications
| Gene ID (Entrez) | 6642 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q13596 |
| Common Name | Human SNX1 (aa 139-155) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.