Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SNX13 Full-length ORF (AAH45667.1, 1 a.a. - 957 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962767 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-767 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-767 Supplier Abnova™ Supplier No. H00023161P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a PHOX domain- and RGS domain-containing protein that belongs to the sorting nexin (SNX) family and the regulator of G protein signaling (RGS) family. The PHOX domain is a phosphoinositide binding domain, and the SNX family members are involved in intracellular trafficking. The RGS family members are regulatory molecules that act as GTPase activating proteins for G alpha subunits of heterotrimeric G proteins. The RGS domain of this protein interacts with G alpha(s), accelerates its GTP hydrolysis, and attenuates G alpha(s)-mediated signaling. Overexpression of this protein delayes lysosomal degradation of the epidermal growth factor receptor. Because of its bifunctional role, this protein may link heterotrimeric G protein signaling and vesicular trafficking. [provided by RefSeq]

Sequence: MLTEASLSIWGWGSLGIVLFLITFGPFVIFYLTFYILCFVGGGLVVTLLFGKTNSEKYLEQCEHSFLPPTSPGVPKCLEEMKREARTIKIDRRLTGANIIDEPLQQVIQFSLRDYVQYWYYTLSDDESFLLEIRQTLQNALIQFATRSKEIDWQPYFTTRIVDDFGTHLRVFRKAQQKITEKDDQVKGTAEDLVDTFFEVEVEMEKEVCRDLVCTSPKDEEGFLRDLCEVLLYLLLPPGDFQNKIMRYFVREILARGILLPLINQLSDPDYINQYVIWMIRDSNCNYEAFMNIIKLSDNIGELEAVRDKAAEELQYLRSLDTAGDDINTIKNQINSLLFVKKVCDSRIQRLQSGKEINTVKLAANFGKLCTVPLDSILVDNVALQFFMDYMQQTGGQAHLFFWMTVEGYRVTAQQQLEVLLSRQRDGKHQTNQTKGLLRAAAVGIYEQYLSEKASPRVTVDDYLVAKLADTLNHEDPTPEIFDDIQRKVYELMLRDERFYPSFRQNALYVRMLAELDMLKDPSFRGSDDGDGESFNGSPTGSINLSLDDLSNVSSDDSVQLHAYISDTGVCNDHGKTYALYAITVHRRNLNSEEMWKTYRRYSDFHDFHMRITEQFESLSSILKLPGKKTFNNMDRDFLEKRKKDLNAYLQLLLAPEMMKASPALAHYVYDFLENKAYSKGKGDFARKMDTFVNPLRNSMRNVSNAVKSLPDNLAEGMTKMSDNMGKMSERLGQDIKQSFFKVPPLIPKTDSDPEHRRVSAQLDDNVDDNIPLRVMLLLMDEVFDLKERNQWLRRNIKNLLQQLIRATYGDTINRKIVDHVDWMTSPEQVADSVKRFRDAFWPNGILAEAVPCRDKSIRMRTRVAGKTKLLAIMPDELKHIIGAETTRKGILRVFEMFQHNQLNRRMVYVFLEGFLETLFPQYKFRELFNKLHSRSKQMQKYKQKLQTTQAPSLQKR

Specifications

Accession Number AAH45667.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23161
Molecular Weight (g/mol) 137.4kDa
Name SNX13 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLTEASLSIWGWGSLGIVLFLITFGPFVIFYLTFYILCFVGGGLVVTLLFGKTNSEKYLEQCEHSFLPPTSPGVPKCLEEMKREARTIKIDRRLTGANIIDEPLQQVIQFSLRDYVQYWYYTLSDDESFLLEIRQTLQNALIQFATRSKEIDWQPYFTTRIVDDFGTHLRVFRKAQQKITEKDDQVKGTAEDLVDTFFEVEVEMEKEVCRDLVCTSPKDEEGFLRDLCEVLLYLLLPPGDFQNKIMRYFVREILARGILLPLINQLSDPDYINQYVIWMIRDSNCNYEAFMNIIKLSDNIGELEAVRDKAAEELQYLRSLDTAGDDINTIKNQINSLLFVKKVCDSRIQRLQSGKEINTVKLAANFGKLCTVPLDSILVDNVALQFFMDYMQQTGGQAHLFFWMTVEGYRVTAQQQLEVLLSRQRDGKHQTNQTKGLLRAAAVGIYEQYLSEKASPRVTVDDYLVAKLADTLNHEDPTPEIFDDIQRKVYELMLRDERFYPSFRQNALYVRMLAELDMLKDPSFRGSDDGDGESFNGSPTGSINLSLDDLSNVSSDDSVQLHAYISDTGVCNDHGKTYALYAITVHRRNLNSEEMWKTYRRYSDFHDFHMRITEQFESLSSILKLPGKKTFNNMDRDFLEKRKKDLNAYLQLLLAPEMMKASPALAHYVYDFLENKAYSKGKGDFARKMDTFVNPLRNSMRNVSNAVKSLPDNLAEGMTKMSDNMGKMSERLGQDIKQSFFKVPPLIPKTDSDPEHRRVSAQLDDNVDDNIPLRVMLLLMDEVFDLKERNQWLRRNIKNLLQQLIRATYGDTINRKIVDHVDWMTSPEQVADSVKRFRDAFWPNGILAEAVPCRDKSIRMRTRVAGKTKLLAIMPDELKHIIGAETTRKGILRVFEMFQHNQLNRRMVYVFLEGFLETLFPQYKFRELFNKLHSRSKQMQKYKQKLQTTQAPSLQKR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0713/RGS-PX1
Common Name SNX13
Gene Symbol SNX13
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.