Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SNX14 Partial ORF (AAH05110, 784 a.a. - 893 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962768 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-768 10 μg
89-962-769 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-768 Supplier Abnova™ Supplier No. H00057231Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. The encoded protein also contains a regulator of G protein signaling (RGS) domain. Regulator of G protein signaling family members are regulatory molecules that act as GTPase activating proteins for G alpha subunits of heterotrimeric G proteins. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: QLFQEHRLVSLITLLRDAIFCENTEPRSLQDKQKGAKQTFEEMMNYIPDLLVKCIGEETKYESIRLLFDGLQQPVLNKQLTYVLLDIVIQELFPELNKVQKEVTSVTSWM

Specifications

Accession Number AAH05110
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57231
Molecular Weight (g/mol) 37.73kDa
Name SNX14 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen QLFQEHRLVSLITLLRDAIFCENTEPRSLQDKQKGAKQTFEEMMNYIPDLLVKCIGEETKYESIRLLFDGLQQPVLNKQLTYVLLDIVIQELFPELNKVQKEVTSVTSWM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC13217/RGS-PX2
Common Name SNX14
Gene Symbol SNX14
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.