Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SNX4 Full-length ORF (NP_003785.1, 1 a.a. - 450 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962786 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-786 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-786 Supplier Abnova™ Supplier No. H00008723P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associated with the long isoform of the leptin receptor and with receptor tyrosine kinases for platelet-derived growth factor, insulin, and epidermal growth factor in cell cultures, but its function is unknown. This protein may form oligomeric complexes with family members. [provided by RefSeq]

Sequence: MEQAPPDPERQLQPAPLEPLGSPDAGLGAAVGKEAEGAGEESSGVDTMTHNNFWLKKIEISVSEAEKRTGRNAMNMQETYTAYLIETRSVEHTDGQSVLTDSLWRRYSEFELLRSYLLVYYPHIVVPPLPEKRAEFVWHKLSADNMDPDFVERRRIGLENFLLRIASHPILCRDKIFYLFLTQEGNWKETVNETGFQLKADSRLKALNATFRVKNPDKRFTDLKHYSDELQSVISHLLRVRARVADRLYGVYKVHGNYGRVFSEWSAIEKEMGDGLQSAGHHMDVYASSIDDILEDEEHYADQLKEYLFYAEALRAVCRKHELMQYDLEMAAQDLASKKQQCEELVTGTVRTFSLKGMTTKLFGQETPEQREARIKVLEEQINEGEQQLKSKNLEGREFVKNAWADIERFKEQKNRDLKEALISYAVMQISMCKKGIQVWTNAKECFSKM

Specifications

Accession Number NP_003785.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8723
Molecular Weight (g/mol) 78.3kDa
Name SNX4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MEQAPPDPERQLQPAPLEPLGSPDAGLGAAVGKEAEGAGEESSGVDTMTHNNFWLKKIEISVSEAEKRTGRNAMNMQETYTAYLIETRSVEHTDGQSVLTDSLWRRYSEFELLRSYLLVYYPHIVVPPLPEKRAEFVWHKLSADNMDPDFVERRRIGLENFLLRIASHPILCRDKIFYLFLTQEGNWKETVNETGFQLKADSRLKALNATFRVKNPDKRFTDLKHYSDELQSVISHLLRVRARVADRLYGVYKVHGNYGRVFSEWSAIEKEMGDGLQSAGHHMDVYASSIDDILEDEEHYADQLKEYLFYAEALRAVCRKHELMQYDLEMAAQDLASKKQQCEELVTGTVRTFSLKGMTTKLFGQETPEQREARIKVLEEQINEGEQQLKSKNLEGREFVKNAWADIERFKEQKNRDLKEALISYAVMQISMCKKGIQVWTNAKECFSKM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name SNX4
Gene Symbol SNX4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.