Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SNX6 Full-length ORF (AAH01798, 1 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962789 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-789 25 μg
89-962-788 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-789 Supplier Abnova™ Supplier No. H00058533P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associates with the long isoform of the leptin receptor, the transforming growth factor-beta family of receptor serine-threonine kinases, and with receptor tyrosine kinases for platelet-derived growth factor, insulin, and epidermal growth factor. This protein may form oligomeric complexes with family member proteins through interactions of both the PX domain and the coiled coil regions of the molecules. Translocation of this protein from the cytoplasm to the nucleus occurs after binding to proviral integration site 1 protein. This gene results in two transcripts encoding two distinct isoforms. [provided by RefSeq]

Sequence: MTKEEFTKMKQELEAEYLAIFKKTVAMHEVFLCRVAAHPILGRDLNFHVFLEYNQDLSVRGKNKKEKLEDFFKNMVKSADGVIVSGVKDVDDFFEHERTFLLEYHNRVKDASAKSDRMTRSHKSAADDYNRIGSSLYALGTQDSTDICKFFLKVSELFDKTRKIEARVSADEDLKLSDLLKYYLRESQAAKDLLYRRSRSLVDYENANKALDKARAKNKDVLQAETSQQLCCQKFEKISESAKQELIDFKTRRVAAFRKNLVELAELELKHAKGNLQLLQNCLAVLNGDT

Specifications

Accession Number AAH01798
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 58533
Molecular Weight (g/mol) 57.64kDa
Name SNX6 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MTKEEFTKMKQELEAEYLAIFKKTVAMHEVFLCRVAAHPILGRDLNFHVFLEYNQDLSVRGKNKKEKLEDFFKNMVKSADGVIVSGVKDVDDFFEHERTFLLEYHNRVKDASAKSDRMTRSHKSAADDYNRIGSSLYALGTQDSTDICKFFLKVSELFDKTRKIEARVSADEDLKLSDLLKYYLRESQAAKDLLYRRSRSLVDYENANKALDKARAKNKDVLQAETSQQLCCQKFEKISESAKQELIDFKTRRVAAFRKNLVELAELELKHAKGNLQLLQNCLAVLNGDT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC3157/MSTP010/TFAF2
Common Name SNX6
Gene Symbol SNX6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.