Learn More
Invitrogen™ Human SOD3 ELISA Kit
Description
Human SOD3 quantitates human SOD3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human SOD3.
Specifications
Specifications
| Accession Number | AI314465; ALS; ALS1; EC SOD; EC-SOD; ECSODPT; extracellular superoxide dismutase; extracellular superoxide dismutase [Cu-Zn]; MGC20077; SOD 3; SOD3; Sod-3; Superoxide dimutase 3; superoxide dismutase; superoxide dismutase 3; superoxide dismutase 3, extracellular; superoxide dismutase B; testicular tissue protein Li 175 |
| Assay Range | 0.65 to 150 ng/mL |
| Assay Sensitivity | 0.65 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 6649 |
| Gene Symbol | SOD3 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.