Learn More
MyBioSource Human ELISA Kits, Factor Related Apoptosis (Fas)
SDP
Supplier:  MyBioSource MBS25333545X96STRIPWEL
Encompass
The MyBioSource Human Fas ELISA Kit quantifies Fas in various biological fluids using a double-antibody sandwich immunoassay. Pre-coated plates bind Fas, which is then detected with biotinylated antibodies and Avidin-HRP, producing a color change measured at 450 nm to determine Fas concentration.
Catalog No. NC2223339
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.