Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Human Soluble gp130 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human gp130/CD130 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), citrate plasma (10 ul).
Specifications
Specifications
| Assay Range | 0.13 to 8 ng/mL (Cell Culture Supernates), 0.25 to 16 ng/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma) |
| Assay Sensitivity | 0.05 ng/mL (Cell Culture Supernates), 0.08 ng/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma) |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Specificity | Natural and Recombinant Human sgp130 |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3572 |
| Sample Volume | Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Citrate Plasma (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
