Learn More
Gibco™ Human Sonic Hedgehog C25II Recombinant Protein, PeproTech®
Description
100-45-1MG will be provided as 2 x 500 μg (100-45-500UG). Recombinant E.coli-derived Human Sonic Hedgehog is a 20.0 kDa protein consisting of 176 amino acid residues, including an N-terminal Ile-Val-Ile sequence substituted for the naturally occurring, chemically modified, Cys residue. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | Q15465 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 6469 |
| Molecular Weight (g/mol) | 20 kDa |
| Name | Human Sonic Hedgehog C25II |
| Purification Method | Purified |
| Quantity | 500 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.