Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SOX10 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF2864SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2864SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2864SP Supplier R&D Systems Supplier No. AF2864SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 32 publications

SOX10 Polyclonal specifically detects SOX10 in Chicken∣Human∣Mouse∣Rat∣Rhesus Macaque∣Transgenic Mouse∣Xenograft samples. It is validated for FACS∣Flow Cytometry∣Immunocytochemistry∣Immunocytochemistry/Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣In Situ∣In Situ Hybridization∣Proximity Ligation Assay (PLA)∣Western Blot.

Specifications

Antigen SOX10
Applications FACS, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Proximity Ligation Assay, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P56693
Gene Alias DOM, MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4, WS4C, WS4mouse, human homolog of
Gene Symbols SOX10
Host Species Goat
Immunogen E. coli-derived recombinant human SOX10 Met1-Ala118 Accession # P56693
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6663
Test Specificity Detects human SOX10 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Chicken, Human, Mouse, Rat, Rhesus Macaque, Transgenic Mouse, Xenograft
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.