Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SOX17 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF1924
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1924 Supplier R&D Systems Supplier No. AF1924

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

SOX17 Polyclonal specifically detects SOX17 in Bovine∣Callithrix jacchus (Common Marmoset)∣Canine∣Chicken∣Chimpanzee∣Cynomolgus Monkey∣Feline∣Human∣Mouse∣Pan troglodytes (Chimpanzee)∣Pig∣Rabbit∣Rat∣Rhesus Macaque∣Transgenic Mouse∣Xenograft samples. It is validated for Bioassay∣ChIP Assay∣Flow Cyomtetry∣Flow Cytomery∣Flow Cytometry∣Functional Assay∣ICC/F∣Immunocytochemistry∣Immunocytochemistry/Immunofluorescence∣Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣Neutralization∣Simple Western∣Western Blot.

Specifications

Antigen SOX17
Applications Bioassay, ChIP Assay, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Simple Western, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 to 2 μg/mL, Simple Western 10 μg/mL, Chromatin Immunoprecipitation (ChIP) 5 μg/5 x 106 cells, Immunocytochemistry 5 to 15 μg/mL
Gene Accession No. Q9H6I2
Gene Alias FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3
Gene Symbols SOX17
Host Species Goat
Immunogen E. coli-derived recombinant human SOX17 Asp177-Val414 Accession # Q9H6I2
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64321
Test Specificity Detects human SOX17 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) SOX18 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Bovine, Marmoset, Canine, Chicken, Chimpanzee, Cynomolgus Monkey, Feline, Human, Mouse, Chimpanzee, Pig, Rabbit, Rat, Rhesus Macaque, Transgenic Mouse, Xenograft
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.