Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SOX17 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. IC1924A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1924A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1924A Supplier R&D Systems Supplier No. IC1924A

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 7 publications

SOX17 Polyclonal specifically detects SOX17 in Human∣Mouse samples. It is validated for Flow Cytometry∣Flow Cytometry (Intracellular Staining)∣Immunocytochemistry∣Immunohistochemistry.

Specifications

Antigen SOX17
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q9H6I2
Gene Alias FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3
Gene Symbols SOX17
Host Species Goat
Immunogen E. coli-derived recombinant human SOX17 Asp177-Val414 Accession # Q9H6I2
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64321
Test Specificity Detects human SOX17 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) SOX2, rhSOX6, and rhSOX15 is observed.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.