Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SOX8 Full-length ORF (NP_055402.2, 1 a.a. - 446 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-892 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-892 Supplier Abnova™ Supplier No. H00030812P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. This protein may be involved in brain development and function. Haploinsufficiency for this protein may contribute to the mental retardation found in haemoglobin H-related mental retardation (ART-16 syndrome). [provided by RefSeq]

Sequence: MLDMSEARSQPPCSPSGTASSMSHVEDSDSDAPPSPAGSEGLGRAGVAVGGARGDPAEAADERFPACIRDAVSQVLKGYDWSLVPMPVRGGGGGALKAKPHVKRPMNAFMVWAQAARRKLADQYPHLHNAELSKTLGKLWRLLSESEKRPFVEEAERLRVQHKKDHPDYKYQPRRRKSAKAGHSDSDSGAELGPHPGGGAVYKAEAGLGDGHHHGDHTGQTHGPPTPPTTPKTELQQAGAKPELKLEGRRPVDSGRQNIDFSNVDISELSSEVMGTMDAFDVHEFDQYLPLGGPAPPEPGQAYGGAYFHAGASPVWAHKSAPSASASPTETGPPRPHIKTEQPSPGHYGDQPRGSPDYGSCSGQSSATPAAPAGPFAGSQGDYGDLQASSYYGAYPGYAPGLYQYPCFHSPRRPYASPLLNGLALPPAHSPTSHWDQPVYTTLTRP

Specifications

Accession Number NP_055402.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 30812
Molecular Weight (g/mol) 73.7kDa
Name SOX8 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLDMSEARSQPPCSPSGTASSMSHVEDSDSDAPPSPAGSEGLGRAGVAVGGARGDPAEAADERFPACIRDAVSQVLKGYDWSLVPMPVRGGGGGALKAKPHVKRPMNAFMVWAQAARRKLADQYPHLHNAELSKTLGKLWRLLSESEKRPFVEEAERLRVQHKKDHPDYKYQPRRRKSAKAGHSDSDSGAELGPHPGGGAVYKAEAGLGDGHHHGDHTGQTHGPPTPPTTPKTELQQAGAKPELKLEGRRPVDSGRQNIDFSNVDISELSSEVMGTMDAFDVHEFDQYLPLGGPAPPEPGQAYGGAYFHAGASPVWAHKSAPSASASPTETGPPRPHIKTEQPSPGHYGDQPRGSPDYGSCSGQSSATPAAPAGPFAGSQGDYGDLQASSYYGAYPGYAPGLYQYPCFHSPRRPYASPLLNGLALPPAHSPTSHWDQPVYTTLTRP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC24837
Common Name SOX8
Gene Symbol SOX8
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.