Learn More
Description
- SFTPD surfactant protein D [ Homo sapiens (human) ]
- Also known as: SP-D; PSP-D; SFTP4; COLEC7
- The SP-D insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- A collectin family (of innate immune modulators) protein consisting of a collagen-like sequence followed by a C-type lectin domain.
- It is constitutively secreted by alveolar lining cells and epithelium associated with tubular structures.
- In addition, SP-D is produced by epithelial cells of the gastrointestinal tract and exocrine glands.
- SP-D binds to patterns of neutral sugars on micro-organisms to mediate direct opsonization and phagocytosis and is an important component of the innate immune system.
- SP-D is part of the innate immune response, protecting the lungs against inhaled microorganisms and chemicals. It may also be involved in surfactant metabolism.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hSFTPD |
| Accession Number | NP_003010 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1141 bp |
| Gene ID (Entrez) | 6441 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
