Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SP2 Partial ORF (NP_003101.2, 71 a.a. - 161 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-911 10 ug
89-962-912 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-911 Supplier Abnova™ Supplier No. H00006668Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the Sp subfamily of Sp/XKLF transcription factors. Sp family proteins are sequence-specific DNA-binding proteins characterized by an amino-terminal trans-activation domain and three carboxy-terminal zinc finger motifs. This protein contains the least conserved DNA-binding domain within the Sp subfamily of proteins, and its DNA sequence specificity differs from the other Sp proteins. It localizes primarily within subnuclear foci associated with the nuclear matrix, and can activate or in some cases repress expression from different promoters. [provided by RefSeq]

Sequence: SPGKNSFGILSSKGNILQIQGSQLSASYPGGQLVFAIQNPTMINKGTRSNANIQYQAVPQIQASNSQTIQVQPNLTNQIQIIPGTNQAIIT

Specifications

Accession Number NP_003101.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6668
Molecular Weight (g/mol) 35.75kDa
Name SP2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen SPGKNSFGILSSKGNILQIQGSQLSASYPGGQLVFAIQNPTMINKGTRSNANIQYQAVPQIQASNSQTIQVQPNLTNQIQIIPGTNQAIIT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name SP2
Gene Symbol SP2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.