Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SP3 Full-length ORF (ADR83384.1, 1 a.a. - 781 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962918 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-918 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-962-918 Supplier Abnova™ Supplier No. H00006670P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. [provided by RefSeq]

Sequence: MTAPEKPVKQEEMAALDVDSGGGGGGGGGHGEYLQQQQQHGNGAVAAAAAAQDTQPSPLALLAATCSKIGPPSPGDDEEEAAAAAGAPAAAGATGDLASAQLGGAPNRWEVLSATPTTIKDEAGNLVQIPSAATSSGQYVLPLQNLQNQQIFSVAPGSDSSNGTVSSVQYQVIPQIQSADGQQVQIGFTGSSDNGGINQESSQIQIIPGSNQTLLASGTPSANIQNLIPQTGQVQVQGVAIGGSSFPGQTQVVANVPLGLPGNITFVPINSVDLDSLGLSGSSQTMTAGINADGHLINTGQAMDSSDNSERTGERVSPDINETNTDTDLFVPTSSSSQLPVTIDSTGILQQNTNSLTTSSGQVHSSDLQGNYIQSPVSEETQAQNIQVSTAQPVVQHLQLQESQQPTSQAQIVQGITPQTIHGVQASGQNISQQALQNLQLQLNPGTFLIQAQTVTPSGQVTWQTFQVQGVQNLQNLQIQNTAAQQITLTPVQTLTLGQVAAGGAFTSTPVSLSTGQLPNLQTVTVNSIDSAGIQLHPGENADSPADIRIKEEEPDPEEWQLSGDSTLNTNDLTHLRVQVVDEEGDQQHQEGKRLRRVACTCPNCKEGGGRGTNLGKKKQHICHIPGCGKVYGKTSHLRAHLRWHSGERPFVCNWMYCGKRFTRSDELQRHRRTHTGEKKFVCPECSKRFMRSDHLAKHIKTHQNKKGIHSSSTVLASVEAARDDTLITAGGTTLILANIQQGSVSGIGTVNTSATSNQDILTNTEIPLQLVTVSGNETME

Specifications

Accession Number ADR83384.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6670
Molecular Weight (g/mol) 112.4kDa
Name SP3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Immunogen MTAPEKPVKQEEMAALDVDSGGGGGGGGGHGEYLQQQQQHGNGAVAAAAAAQDTQPSPLALLAATCSKIGPPSPGDDEEEAAAAAGAPAAAGATGDLASAQLGGAPNRWEVLSATPTTIKDEAGNLVQIPSAATSSGQYVLPLQNLQNQQIFSVAPGSDSSNGTVSSVQYQVIPQIQSADGQQVQIGFTGSSDNGGINQESSQIQIIPGSNQTLLASGTPSANIQNLIPQTGQVQVQGVAIGGSSFPGQTQVVANVPLGLPGNITFVPINSVDLDSLGLSGSSQTMTAGINADGHLINTGQAMDSSDNSERTGERVSPDINETNTDTDLFVPTSSSSQLPVTIDSTGILQQNTNSLTTSSGQVHSSDLQGNYIQSPVSEETQAQNIQVSTAQPVVQHLQLQESQQPTSQAQIVQGITPQTIHGVQASGQNISQQALQNLQLQLNPGTFLIQAQTVTPSGQVTWQTFQVQGVQNLQNLQIQNTAAQQITLTPVQTLTLGQVAAGGAFTSTPVSLSTGQLPNLQTVTVNSIDSAGIQLHPGENADSPADIRIKEEEPDPEEWQLSGDSTLNTNDLTHLRVQVVDEEGDQQHQEGKRLRRVACTCPNCKEGGGRGTNLGKKKQHICHIPGCGKVYGKTSHLRAHLRWHSGERPFVCNWMYCGKRFTRSDELQRHRRTHTGEKKFVCPECSKRFMRSDHLAKHIKTHQNKKGIHSSSTVLASVEAARDDTLITAGGTTLILANIQQGSVSGIGTVNTSATSNQDILTNTEIPLQLVTVSGNETME
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686O1631/SPR-2
Common Name SP3
Gene Symbol SP3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.