Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SPACA1 Full-length ORF (NP_112222.1, 1 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962932 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-932 25 μg
89-962-931 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-932 Supplier Abnova™ Supplier No. H00081833P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The correlation of anti-sperm antibodies with cases of unexplained infertility implicates a role for these antibodies in blocking fertilization. Improved diagnosis and treatment of immunologic infertility, as well as identification of proteins for targeted contraception, are dependent on the identification and characterization of relevant sperm antigens. The protein expressed by this gene is recognized by anti-sperm antibodies from infertile males. Furthermore, antibodies generated against the recombinant protein block in vitro fertilization. This protein localizes to the acrosomal membrane of spermatids and mature spermatozoa where it is thought to play a role in acrosomal morphogenesis and in sperm-egg binding and fusion, respectively. [provided by RefSeq]

Sequence: MSPRGTGCSAGLLMTVGWLLLAGLQSARGTNVTAAVQDAGLAHEGEGEEETENNDSETAENYAPPETEDVSNRNVVKEVEFGMCTVTCGIGVREVILTNGCPGGESKCVVRVEECRGPTDCGWGKPISESLESVRLACIHTSPLNRFKYMWKLLRQDQQSIILVNDSAILEVRKESHPLAFECDTLDNNEIVATIKFTVYTSSELQMRRSSLPATDAALIFVLTIGVIICVFIIFLLIFIIINWAAVKAFWGAKASTPEVQSEQSSVRYKDSTSLDQLPTEMPGEDDALSEWNE

Specifications

Accession Number NP_112222.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 81833
Molecular Weight (g/mol) 58.5kDa
Name SPACA1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MSPRGTGCSAGLLMTVGWLLLAGLQSARGTNVTAAVQDAGLAHEGEGEEETENNDSETAENYAPPETEDVSNRNVVKEVEFGMCTVTCGIGVREVILTNGCPGGESKCVVRVEECRGPTDCGWGKPISESLESVRLACIHTSPLNRFKYMWKLLRQDQQSIILVNDSAILEVRKESHPLAFECDTLDNNEIVATIKFTVYTSSELQMRRSSLPATDAALIFVLTIGVIICVFIIFLLIFIIINWAAVKAFWGAKASTPEVQSEQSSVRYKDSTSLDQLPTEMPGEDDALSEWNE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC32952/SAMP32
Common Name SPACA1
Gene Symbol SPACA1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.