Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SPANXB1 Full-length ORF (AAH34472, 1 a.a. - 103 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962955 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-955 10 μg
89-962-956 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-955 Supplier Abnova™ Supplier No. H00728695P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family of cancer/testis-associated genes, which are located in a cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene maps to chromosome X in a head-to-tail orientation with SPANX family member B2, which appears to be a duplication of the B1 locus. The SPANXB genes are unique members of this gene family, since they contain an additional 18 nt in their coding region compared to the majority of family members. Although the protein encoded by this gene contains consensus nuclear localization signals, the major site for subcellular localization of expressed protein is in the cytoplasmic droplets of ejaculated spermatozoa. This protein provides a biochemical marker for studying the unique structures in spermatazoa, while attempting to further define its role in spermatogenesis. [provided by RefSeq]

Sequence: MGQQSSVRRLKRSVPCESNEANEANEANKTMPETPTGDSDPQPAPKKMKTSESSTILVVRYRRNVKRTSPEELVNDHARENRINPDQMEEEEFIEITTERPKK

Specifications

Accession Number AAH34472
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 728695
Molecular Weight (g/mol) 37.07kDa
Name SPANXB1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MGQQSSVRRLKRSVPCESNEANEANEANKTMPETPTGDSDPQPAPKKMKTSESSTILVVRYRRNVKRTSPEELVNDHARENRINPDQMEEEEFIEITTERPKK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias B1/SPANX-B/SPANXB
Common Name SPANXB1
Gene Symbol SPANXB1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.