Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human SPARC Quantikine ELISA Kit
SDP

Catalog No. DSP00 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DSP00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DSP00 Supplier R&D Systems™ Supplier No. DSP00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human SPARC Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human SPARC Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human SPARC in cell culture supernates (10 ul), serum (10 ul), platelet-poor edta plasma (20 ul), platelet-poor heparin plasma (20 ul), human milk (50 ul).

Specifications

Assay Range 1.56 - 50ng/mL
Assay Sensitivity 0.269ng/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Human Milk, Platelet-poor Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human SPARC
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 6678
Sample Volume Cell Culture Supernates (10uL),Serum (10uL),Platelet-poor EDTA Plasma (20uL),Platelet-poor Heparin Plasma (20uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.