Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SPG21 Full-length ORF (NP_057714.1, 1 a.a. - 308 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963025 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-025 25 μg
89-963-024 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-025 Supplier Abnova™ Supplier No. H00051324P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene was identified by a two-hybrid screen using CD4 as the bait. It binds to the hydrophobic C-terminal amino acids of CD4 which are involved in repression of T cell activation. The interaction with CD4 is mediated by the noncatalytic alpha/beta hydrolase fold domain of this protein. It is thus proposed that this gene product modulates the stimulatory activity of CD4. At least three different transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MGEIKVSPDYNWFRGTVPLKKIIVDDDDSKIWSLYDAGPRSIRCPLIFLPPVSGTADVFFRQILALTGWGYRVIALQYPVYWDHLEFCDGFRKLLDHLQLDKVHLFGASLGGFLAQKFAEYTHKSPRVHSLILCNSFSDTSIFNQTWTANSFWLMPAFMLKKIVLGNFSSGPVDPMMADAIDFMVDRLESLGQSELASRLTLNCQNSYVEPHKIRDIPVTIMDVFDQSALSTEAKEEMYKLYPNARRAHLKTGGNFPYLCRSAEVNLYVQIHLLQFHGTKYAAIDPSMVSAEELEVQKGSLGISQEEQ

Specifications

Accession Number NP_057714.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51324
Molecular Weight (g/mol) 61.4kDa
Name SPG21 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MGEIKVSPDYNWFRGTVPLKKIIVDDDDSKIWSLYDAGPRSIRCPLIFLPPVSGTADVFFRQILALTGWGYRVIALQYPVYWDHLEFCDGFRKLLDHLQLDKVHLFGASLGGFLAQKFAEYTHKSPRVHSLILCNSFSDTSIFNQTWTANSFWLMPAFMLKKIVLGNFSSGPVDPMMADAIDFMVDRLESLGQSELASRLTLNCQNSYVEPHKIRDIPVTIMDVFDQSALSTEAKEEMYKLYPNARRAHLKTGGNFPYLCRSAEVNLYVQIHLLQFHGTKYAAIDPSMVSAEELEVQKGSLGISQEEQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ACP33/BM-019/GL010/MASPARDIN/MAST
Common Name SPG21
Gene Symbol SPG21
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.