Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SPTLC2 Partial ORF (NP_004854, 453 a.a. - 561 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963151 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-151 10 μg
89-963-152 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-151 Supplier Abnova™ Supplier No. H00009517Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a long chain base subunit of serine palmitoyltransferase. Serine palmitoyltransferase, which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine. Mutations in this gene were identified in patients with hereditary sensory neuropathy type I. Alternatively spliced variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: LKEMGFIIYGNEDSPVVPLMLYMPAKIGAFGREMLKRNIGVVVVGFPATPIIESRARFCLSAAHTKEILDTALKEIDEVGDLLQLKYSRHRLVPLLDRPFDETTYEETE

Specifications

Accession Number NP_004854
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9517
Molecular Weight (g/mol) 37.73kDa
Name SPTLC2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen LKEMGFIIYGNEDSPVVPLMLYMPAKIGAFGREMLKRNIGVVVVGFPATPIIESRARFCLSAAHTKEILDTALKEIDEVGDLLQLKYSRHRLVPLLDRPFDETTYEETE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0526/LCB2/SPT2
Common Name SPTLC2
Gene Symbol SPTLC2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.