Learn More
Description
- MSR1 macrophage scavenger receptor 1 [ Homo sapiens (human) ]
- Also known as: SRA; SR-A; CD204; SRAI; phSR1; phSR2; SCARA1; SR-AII; SRAIII
- The scavenger receptor (SR) family is a group of membrane receptors that share the common ability to bind and internalize modified forms of low-density lipoproteins (mLDL).
- The three SR-A alternatively spliced isoforms (I, II, and III) and a structurally similar protein, MARCO, make up the A class of homotrimeric, type II membrane proteins.
- SR-AI has an N-terminal cytoplasmic domain, transmembrane domain, spacer domain, a-helical coiled coil domain, collagen-like domain, and C-terminal cysteine-rich domain.
- SR-As are expressed by most tissue macrophages and dendritic cells.
- SR-As bind a broad range of polyanionic ligands and therefore have been implicated in many physiological and pathophysiological functions.
- The SR-AI/MSR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hMSR1 |
| Accession Number | NP_619729 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1369 bp |
| Gene ID (Entrez) | 4481 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
