Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SR-AI/MSR1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7065254
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB27081SP 25 μg
MAB27081 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB27081SP Supplier R&D Systems Supplier No. MAB27081SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

SR-AI/MSR Monoclonal specifically detects SR-AI/MSR in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.

Specifications

Antigen SR-AI/MSR
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 351620
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P21757
Gene Alias CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, MSR1, phSR1, phSR2, SCARA1, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A, SRAI
Gene Symbols MSR1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SR-AI Lys77-Leu451 Accession # P21757
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4481
Test Specificity Detects human SR-AI in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse SR-AI.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.