Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SREC-II/SCARF2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2527
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2527 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2527 Supplier R&D Systems Supplier No. BAF2527
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SREC-II/SCARF2 Polyclonal specifically detects SREC-II/SCARF2 in Human samples. It is validated for Western Blot.

Specifications

Antigen SREC-II/SCARF2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q96GP6
Gene Alias HUMZD58C02, SCARF2, scavenger receptor class F, member 2, Scavenger receptor expressed by endothelial cells 2 protein, SREC2NSR1, SRECII, SREC-IIscavenger receptor class F member 2, SRECRP-1, SREPCR
Gene Symbols SCARF2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human SREC-II/SCARF2 Ala42-Gly442 Accession # Q96GP6
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91179
Test Specificity Detects human SREC-II/SCARF2 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human SREC-I is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.