Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SRPK1 Full-length ORF (AAH38292.1, 1 a.a. - 655 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963211 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-211 10 ug
89-963-212 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-211 Supplier Abnova™ Supplier No. H00006732P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, KA, WB-Re

This gene encodes a serine/arginine protein kinase specific for the SR (serine/arginine-rich domain) family of splicing factors. The protein localizes to the nucleus and the cytoplasm. It is thought to play a role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. A second alternatively spliced transcript variant for this gene has been described, but its full length nature has not been determined. [provided by RefSeq]

Sequence: MERKVLALQARKKRTKAKKDKAQRKSETQHRGSAPHSESDLPEQEEEILGSDDDEQEDPNDYCKGGYHLVKIGDLFNGRYHVIRKLGWGHFSTVWLSWDIQGKKFVAMKVVKSAEHYTETALDEIRLLKSVRNSDPNDPNREMVVQLLDDFKISGVNGTHICMVFEVLGHHLLKWIIKSNYQGLPLPCVKKIIQQVLQGLDYLHTKCRITHTDIKPENILLSVNEQYIRRLAAEATEWQRSGAPPPSGSAVSTAPQPKPADKMSKNKKKKLKKKQKRQAELLEKRMQEIEEMEKESGPGQKRPNKQEESESPVERPLKENPPNKMTQEKLEESSTIGQDQTLMERDTEGGAAEINCNGVIEVINYTQNSNNETLRHKEDLHNANDCDVQNLNQESSFLSSQNGDSSTSQETDSCTPITSEVSDTMVCQSSSTVGQSFSEQHISQLQESIRAEIPCEDEQEQEHNGPLDNKGKSTAGNFLVNPLEPKNAEKLKVKIADLGNACWVHKHFTEDIQTRQYRSLEVLIGSGYNTPADIWSTACMAFELATGDYLFEPHSGEEYTRDEDHIALIIELLGKVPRKLIVAGKYSKEFFTKKGDLKHITKLKPWGLFEVLVEKYEWSQEEAAGFTDFLLPMLELIPEKRATAAECLRHPWLNS

Specifications

Accession Number AAH38292.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Kinase Assay, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6732
Molecular Weight (g/mol) 97.79kDa
Name SRPK1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MERKVLALQARKKRTKAKKDKAQRKSETQHRGSAPHSESDLPEQEEEILGSDDDEQEDPNDYCKGGYHLVKIGDLFNGRYHVIRKLGWGHFSTVWLSWDIQGKKFVAMKVVKSAEHYTETALDEIRLLKSVRNSDPNDPNREMVVQLLDDFKISGVNGTHICMVFEVLGHHLLKWIIKSNYQGLPLPCVKKIIQQVLQGLDYLHTKCRITHTDIKPENILLSVNEQYIRRLAAEATEWQRSGAPPPSGSAVSTAPQPKPADKMSKNKKKKLKKKQKRQAELLEKRMQEIEEMEKESGPGQKRPNKQEESESPVERPLKENPPNKMTQEKLEESSTIGQDQTLMERDTEGGAAEINCNGVIEVINYTQNSNNETLRHKEDLHNANDCDVQNLNQESSFLSSQNGDSSTSQETDSCTPITSEVSDTMVCQSSSTVGQSFSEQHISQLQESIRAEIPCEDEQEQEHNGPLDNKGKSTAGNFLVNPLEPKNAEKLKVKIADLGNACWVHKHFTEDIQTRQYRSLEVLIGSGYNTPADIWSTACMAFELATGDYLFEPHSGEEYTRDEDHIALIIELLGKVPRKLIVAGKYSKEFFTKKGDLKHITKLKPWGLFEVLVEKYEWSQEEAAGFTDFLLPMLELIPEKRATAAECLRHPWLNS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias SFRSK1
Common Name SRPK1
Gene Symbol SRPK1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.