Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SRPK3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7230SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7230SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7230SP Supplier R&D Systems Supplier No. MAB7230SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SRPK3/STK23 Monoclonal specifically detects SRPK3/STK23 in Human samples. It is validated for Western Blot.

Specifications

Antigen SRPK3/STK23
Applications Western Blot
Classification Monoclonal
Clone 722534
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UPE1
Gene Alias MGC102944, MSSK1, MSSK-1, MSSK1EC 2.7.11.1, Muscle-specific serine kinase 1, serine arginine rich protein-specific kinase 3, Serine/arginine-rich protein-specific kinase 3, serine/threonine kinase 23, Serine/threonine-protein kinase 23, serine/threonine-protein kinase SRPK3, SFRS protein kinase 3, SR-protein-specific kinase 3, SRSF protein kinase 3, STK23
Gene Symbols SRPK3
Host Species Mouse
Immunogen E.coli-derived recombinant human SRPK3 Arg247-Ser316 Accession # Q9UPE1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26576
Test Specificity Detects human SRPK3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) SRPK1 or rhSRPK2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.