Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SRRM1 Partial ORF (NP_005830.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963245 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-245 10 μg
89-963-246 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-245 Supplier Abnova™ Supplier No. H00010250Q01S

Used for AP, Array, ELISA, WB-Re

Sequence: MDAGFFRGTSAEQDNRFSNKQKKLLKQLKFAECLEKKVDMSKVNLEVIKPWITKRVTEILGFEDDVVIEFIFNQLEVKNPDSKMMQINLTGFLNGKNARE

Specifications

Accession Number NP_005830.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10250
Molecular Weight (g/mol) 36.74kDa
Name SRRM1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MDAGFFRGTSAEQDNRFSNKQKKLLKQLKFAECLEKKVDMSKVNLEVIKPWITKRVTEILGFEDDVVIEFIFNQLEVKNPDSKMMQINLTGFLNGKNARE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 160-KD/MGC39488/POP101/SRM160
Common Name SRRM1
Gene Symbol SRRM1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.