Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ST3GAL1 ELISA Kit

Catalog No. EH438RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH438RBX5 5 x 96 Tests
EH438RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH438RBX5 Supplier Invitrogen™ Supplier No. EH438RBX5
Add to Cart
Add to Cart

ELISA

Human ST3GAL1 quantitates human ST3GAL1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ST3GAL1.

ST3GAL1 is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. It is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of ST3GAL1 protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B.The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of the encoded protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B. Two transcript variants encoding the same protein have been found for this gene. Other transcript variants may exist, but have not been fully characterized yet.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 5330418N22Rik; AI467004; Alpha 2,3-ST 1; beta-galactoside alpha-2,3-sialyltransferase 1; CMP-N-acetylneuraminate: [beta-galactosidase alpha-2,3 ] sialytransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1; Gal beta1,3 GalNAc alpha2,3 sialyltransferase; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; MGC9183; Sialyltransferase 4 A; sialyltransferase 4 A (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4 A (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 4 A (beta-galactoside alpha-2,3-sialytransferase); SIAT4; SIAT4A; SIAT4-A; SIATFL; ST3 beta-galactoside alpha-2,3-sialyltransferase 1; ST3Gal I; St3gal1; St3gal-1; ST3GalA; ST3GalA0.1; ST3GalI; ST3GAL-I sialyltransferase; ST3GalIA; ST3GalIA,1; ST3O
Assay Range 1.8 to 400 ng/mL
Assay Sensitivity 1.8 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 6482
Gene Symbol ST3GAL1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target ST3GAL1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.