Learn More
Description
Specifications
Specifications
| Gene | ST3GAL3 |
| Accession Number | NP_006270 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl |
| Insert Type | 1141bp |
| pH | 8.5 |
| Gene ID (Entrez) | 6487 |
| Gene Alias | alpha 2,3-sialyltransferase III, Alpha 2,3-ST 3, alpha-2,3-sialyltransferase II, Beta-galactoside alpha-2,3-sialyltransferase 3, CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase, EC 2.4.99.6, Gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase, Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase, MRT12, N-acetyllactosaminide alpha-2,3-sialyltransferase, Sialyltransferase 6, Siat3, SIAT6, SIAT6sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase), ST3 beta-Gal alpha-2,3-Sialyltransferase 3, ST3 beta-galactoside alpha-2,3-sialyltransferase 3, ST3Gal III, ST3GALII, St3Gal-III, ST3GalIII, ST3N |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
