Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ST3GAL5 Full-length ORF (AAH65936.1, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-371 10 μg
89-963-372 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-371 Supplier Abnova™ Supplier No. H00008869P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintenance of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MPSEYTYVKLRSDCSRPSLQWYTRAQSKMRRPSLLLKDILKCTLLVFGVWILYILKLNYTTEECDMKKMHYVDPDRVKRAQKYAQQVLQKECRPKFAKTSMALLFEHRYSVDLLPFVQKAPKDSEAESKYDPPFGFRKFSSKVQTLLELLPEHDLPEHLKAKTCRRCVVIGSGGILHGLELGHTLNQFDVVIRLNSAPVEGYSEHVGNKTTIRMTYPEGAPLSDLEYYSNDLFVAVLFKSVDFNWLQAMVKKETLPFWVRLFFWKQVAEKIPLQPKHFRILNPVIIKETAFDILQYSEPQSRFWGRDKNVPTIGVIAVVLATHLCDEVSLAGFGYDLNQPRTPLHYFDSQCMAAMNFQTMHNVTTETKFLLKLVKEGVVKDLSGGIDREF

Specifications

Accession Number AAH65936.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8869
Molecular Weight (g/mol) 71.5kDa
Name ST3GAL5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MPSEYTYVKLRSDCSRPSLQWYTRAQSKMRRPSLLLKDILKCTLLVFGVWILYILKLNYTTEECDMKKMHYVDPDRVKRAQKYAQQVLQKECRPKFAKTSMALLFEHRYSVDLLPFVQKAPKDSEAESKYDPPFGFRKFSSKVQTLLELLPEHDLPEHLKAKTCRRCVVIGSGGILHGLELGHTLNQFDVVIRLNSAPVEGYSEHVGNKTTIRMTYPEGAPLSDLEYYSNDLFVAVLFKSVDFNWLQAMVKKETLPFWVRLFFWKQVAEKIPLQPKHFRILNPVIIKETAFDILQYSEPQSRFWGRDKNVPTIGVIAVVLATHLCDEVSLAGFGYDLNQPRTPLHYFDSQCMAAMNFQTMHNVTTETKFLLKLVKEGVVKDLSGGIDREF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias SIAT9/SIATGM3S/ST3GalV
Common Name ST3GAL5
Gene Symbol ST3GAL5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.