Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ST6GAL2 Partial ORF (NP_115917.1, 33 a.a. - 141 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963381 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-381 10 μg
89-963-382 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-381 Supplier Abnova™ Supplier No. H00084620Q01S

Used for AP, Array, ELISA, WB-Re

Sialyltransferases, such as ST6GAL2 (EC 2.4.99.1), are type II transmembrane proteins that catalyze the transfer of sialic acid from CMP-sialic acid to an acceptor carbohydrate, usually to the terminal ends of carbohydrate chains.[supplied by OMIM]

Sequence: SNPAEPVPSSLSFLETRRLLPVQGKQRAIMGAAHEPSPPGGLDARQALPRAHPAGSFHAGPGDLQKWAQSQDGFEHKEFFSSQVGRKSQSAFYPEDDDYFFAAGQPGWH

Specifications

Accession Number NP_115917.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84620
Molecular Weight (g/mol) 37.73kDa
Name ST6GAL2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen SNPAEPVPSSLSFLETRRLLPVQGKQRAIMGAAHEPSPPGGLDARQALPRAHPAGSFHAGPGDLQKWAQSQDGFEHKEFFSSQVGRKSQSAFYPEDDDYFFAAGQPGWH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ30711/FLJ37730/FLJ38334/KIAA1877/SIAT2/ST6GalII
Common Name ST6GAL2
Gene Symbol ST6GAL2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.