Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ST7L Full-length ORF (NP_060214.2, 1 a.a. - 575 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963403 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-403 10 μg
89-963-404 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-403 Supplier Abnova™ Supplier No. H00054879P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene was identified by its similarity to the ST7 tumor suppressor gene found in the chromosome 7q31 region. This gene is clustered in a tail-to-tail manner with the WNT2B gene in a chromosomal region known to be deleted and rearranged in a variety of cancers. The function of this gene has not yet been determined. Several transcript variants encoding many different isoforms have been described, but some have not been fully characterized. [provided by RefSeq]

Sequence: MADRGGVGEAAAVGASPASVPGLNPTLGWRERLRAGLAGTGASLWFVAGLGLLYALRIPLRLCENLAAVTVFLNSLTPKFYVALTGTSSLISGLIFIFEWWYFHKHGTSFIEQVSVSHLQPLMGGTESSISEPGSPSRNRENETSRQNLSECKVWRNPLNLFRGAEYRRYTWVTGKEPLTYYDMNLSAQDHQTFFTCDTDFLRPSDTVMQKAWRERNPPARIKAAYQALELNNDCATAYVLLAEEEATTIVDAERLFKQALKAGETIYRQSQQCQHQSPQHEAQLRRDTNVLVYIKRRLAMCARKLGRIREAVKIMRDLMKEFPPLTMLNIHENLLESLLELQAYPDVQAVLAKYDDISLPKSAAICYTAALLKTRTVSEKFSPETASRRGLSTAEINAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLQHWKRIEGALNLLQCTWEGTFRMIPYPLEKGHLFYPYPSCTETADRELLPTFHHVSVYPKKELPLFIHFTAGFCSSTAMIAILTHQFPEIMGIFAKAVLGLWCPQPWASSGFEENTQDLKSEDLGLSSG

Specifications

Accession Number NP_060214.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54879
Molecular Weight (g/mol) 91.2kDa
Name ST7L (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MADRGGVGEAAAVGASPASVPGLNPTLGWRERLRAGLAGTGASLWFVAGLGLLYALRIPLRLCENLAAVTVFLNSLTPKFYVALTGTSSLISGLIFIFEWWYFHKHGTSFIEQVSVSHLQPLMGGTESSISEPGSPSRNRENETSRQNLSECKVWRNPLNLFRGAEYRRYTWVTGKEPLTYYDMNLSAQDHQTFFTCDTDFLRPSDTVMQKAWRERNPPARIKAAYQALELNNDCATAYVLLAEEEATTIVDAERLFKQALKAGETIYRQSQQCQHQSPQHEAQLRRDTNVLVYIKRRLAMCARKLGRIREAVKIMRDLMKEFPPLTMLNIHENLLESLLELQAYPDVQAVLAKYDDISLPKSAAICYTAALLKTRTVSEKFSPETASRRGLSTAEINAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLQHWKRIEGALNLLQCTWEGTFRMIPYPLEKGHLFYPYPSCTETADRELLPTFHHVSVYPKKELPLFIHFTAGFCSSTAMIAILTHQFPEIMGIFAKAVLGLWCPQPWASSGFEENTQDLKSEDLGLSSG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FAM4B/FLJ11657/FLJ20284/ST7R/STLR
Common Name ST7L
Gene Symbol ST7L
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.