Learn More
Abnova™ Human ST7L Full-length ORF (NP_060214.2, 1 a.a. - 575 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene was identified by its similarity to the ST7 tumor suppressor gene found in the chromosome 7q31 region. This gene is clustered in a tail-to-tail manner with the WNT2B gene in a chromosomal region known to be deleted and rearranged in a variety of cancers. The function of this gene has not yet been determined. Several transcript variants encoding many different isoforms have been described, but some have not been fully characterized. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_060214.2 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 54879 |
| Molecular Weight (g/mol) | 91.2kDa |
| Name | ST7L (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MADRGGVGEAAAVGASPASVPGLNPTLGWRERLRAGLAGTGASLWFVAGLGLLYALRIPLRLCENLAAVTVFLNSLTPKFYVALTGTSSLISGLIFIFEWWYFHKHGTSFIEQVSVSHLQPLMGGTESSISEPGSPSRNRENETSRQNLSECKVWRNPLNLFRGAEYRRYTWVTGKEPLTYYDMNLSAQDHQTFFTCDTDFLRPSDTVMQKAWRERNPPARIKAAYQALELNNDCATAYVLLAEEEATTIVDAERLFKQALKAGETIYRQSQQCQHQSPQHEAQLRRDTNVLVYIKRRLAMCARKLGRIREAVKIMRDLMKEFPPLTMLNIHENLLESLLELQAYPDVQAVLAKYDDISLPKSAAICYTAALLKTRTVSEKFSPETASRRGLSTAEINAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLQHWKRIEGALNLLQCTWEGTFRMIPYPLEKGHLFYPYPSCTETADRELLPTFHHVSVYPKKELPLFIHFTAGFCSSTAMIAILTHQFPEIMGIFAKAVLGLWCPQPWASSGFEENTQDLKSEDLGLSSG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.