Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ST8SIA2 Partial ORF (NP_006002, 40 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963407 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-407 10 μg
89-963-408 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-407 Supplier Abnova™ Supplier No. H00008128Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29. [provided by RefSeq]

Sequence: TIRSAVNSLHSKSNRAEVVINGSSSPAVVDRSNESIKHNIQPASSKWRHNQTLSLRIRKQILKFLDAEKDISVLKGTLKPGDIIHYIFDRDSTMNVSQNLYELLPRT

Specifications

Accession Number NP_006002
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8128
Molecular Weight (g/mol) 37.51kDa
Name ST8SIA2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen TIRSAVNSLHSKSNRAEVVINGSSSPAVVDRSNESIKHNIQPASSKWRHNQTLSLRIRKQILKFLDAEKDISVLKGTLKPGDIIHYIFDRDSTMNVSQNLYELLPRT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HsT19690/MGC116854/MGC116857/SIAT8B/ST8SIA-II/STX
Common Name ST8SIA2
Gene Symbol ST8SIA2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.