Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human STAG1 Partial ORF (AAH64699, 1112 a.a. - 1221 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963423 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-423 25 μg
89-963-422 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-423 Supplier Abnova™ Supplier No. H00010274Q01L

Used for AP, Array, ELISA, WB-Re

This gene is a member of the SCC3 family and is expressed in the nucleus. It encodes a component of cohesin, a multisubunit protein complex that provides sister chromatid cohesion along the length of a chromosome from DNA replication through prophase and prometaphase, after which it is dissociated in preparation for segregation during anaphase. [provided by RefSeq]

Sequence: LPAPQLTSTVLRENSRPMGDQIQEPESEHGSEPDFLHNRGLMEEDAEPIFEDVMMSSRSQLEDMNEEFEDTMVIDLPPSRNRRERAELRPDFFDSAAIIEDDSGFGMPMF

Specifications

Accession Number AAH64699
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10274
Molecular Weight (g/mol) 37.73kDa
Name STAG1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen LPAPQLTSTVLRENSRPMGDQIQEPESEHGSEPDFLHNRGLMEEDAEPIFEDVMMSSRSQLEDMNEEFEDTMVIDLPPSRNRRERAELRPDFFDSAAIIEDDSGFGMPMF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp781D1416/SA1
Common Name STAG1
Gene Symbol STAG1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.