Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human STARD4 Full-length ORF (NP_631903.1, 1 a.a. - 205 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963448 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-448 10 μg
89-963-449 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-448 Supplier Abnova™ Supplier No. H00134429P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Cholesterol homeostasis is regulated, at least in part, by sterol regulatory element (SRE)-binding proteins (e.g., SREBP1; MIM 184756) and by liver X receptors (e.g., LXRA; MIM 602423). Upon sterol depletion, LXRs are inactive and SREBPs are cleaved, after which they bind promoter SREs and activate genes involved in cholesterol biosynthesis and uptake. Sterol transport is mediated by vesicles or by soluble protein carriers, such as steroidogenic acute regulatory protein (STAR; MIM 600617). STAR is homologous to a family of proteins containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain, including STARD4 (Soccio et al., 2002 [PubMed 12011452]).[supplied by OMIM]

Sequence: MEGLSDVASFATKLKNTLIQYHSIEEDKWRVAKKTKDVTVWRKPSEEFNGYLYKAQGVIDDLVYSIIDHIRPGPCRLDWDSLMTSLDILENFEENCCVMRYTTAGQLWNIISPREFVDFSYTVGYKEGLLSCGISLDWDEKRPEFVRGYNHPCGWFCVPLKDNPNQSLLTGYIQTDLRGMIPQSAVDTAMASTLTNFYGDLRKAL

Specifications

Accession Number NP_631903.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 134429
Molecular Weight (g/mol) 49.9kDa
Name STARD4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEGLSDVASFATKLKNTLIQYHSIEEDKWRVAKKTKDVTVWRKPSEEFNGYLYKAQGVIDDLVYSIIDHIRPGPCRLDWDSLMTSLDILENFEENCCVMRYTTAGQLWNIISPREFVDFSYTVGYKEGLLSCGISLDWDEKRPEFVRGYNHPCGWFCVPLKDNPNQSLLTGYIQTDLRGMIPQSAVDTAMASTLTNFYGDLRKAL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name STARD4
Gene Symbol STARD4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.